missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA 8270 Surrogate Standard, Certified Reference Material, 4,000 μg/mL Each Component in Dichloromethane, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 4000 μg/mL each component in dichloromethane |
| Synonym | EPA 8270 Surrogate Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 2-Fluorobiphenyl; 2-Fluorophenol; Nitrobenzene-d5; Phenol-d5; p-Terphenyl-d14; 2,4,6-Tribromophenol |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC