missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA 8260 Calibration Mix 2, Analytical Standard, 2,000 μg/mL Each Component in Methanol: Water (19:1), MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Concentration | 2,000 μg/mL each component in methanol: water (19:1) |
| Type | Organic Pollutant Standard |
| Quantity | 1.5 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Acetone; 2-Butanone; Carbon disulfide; 2-Chloroethyl vinyl ether; 2-Hexanone; Iodomethane; 4-Methyl-2-pentanone; Vinyl acetate |