missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EP Color Standard B7, Brown Standard Solution, Ricca Chemical

$553.24 - $553.24

Chemical Identifiers

CAS 7732-18-5,7647-01-0,7791-13-1,7758-99-8,10025-77-1
CAS Min % 94.0,1.4,0.86,0.72,0.67
CAS Max % 98.0,1.45,0.9,0.75,0.7
Physical Form Liquid
Chemical Name or Material EP Color Standard B, Brown Standard Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2880-4 SDS Ricca Chemical Company
RI28804
Glass amber 120mL Each for $553.24
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,7791-13-1,7758-99-8,10025-77-1
98.0,1.45,0.9,0.75,0.7
EP Color Standard B, Brown Standard Solution
94.0,1.4,0.86,0.72,0.67
Liquid

Specifications

7732-18-5,7647-01-0,7791-13-1,7758-99-8,10025-77-1
98.0,1.45,0.9,0.75,0.7
Glass amber
120mL
94.0,1.4,0.86,0.72,0.67
Liquid
Pale amber
EP Color Standard B, Brown Standard Solution