missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eosin Y Stain, 0.25% (w/v) in 57% (v/v) Alcohol, Acidified Working Solution, Ricca Chemical
$145.41 - $145.41
Specifications
Chemical Name or Material | Eosin Y Stain |
---|---|
Name Note | Acidified Working Solution |
CAS Min % | 66.0,28.79,1.86,0.56,0.5 |
CAS Max % | 69.0,29.97,1.94,0.59,0.52 |
CAS | 64-17-5,67-63-0,64-19-7,108-10-1,17372-87-1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
2845-16
|
Ricca Chemical Company
284516 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
Specifications
Eosin Y Stain | |
66.0,28.79,1.86,0.56,0.5 | |
64-17-5,67-63-0,64-19-7,108-10-1,17372-87-1 | |
Natural Poly Bottle | |
Orange/Red |
Acidified Working Solution | |
69.0,29.97,1.94,0.59,0.52 | |
0.25% (w/v) | |
57% (v/v) Alcohol | |
Liquid |