Please login to your online account to display your discounted pricing

Eosin Y, Reagent Grade, LabChem™

$82.17 - $82.17

Chemical Identifiers

Chemical Name or Material Eosin Y
CAS 17372-87-1
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C20H6Br4O5·2Na
Formula Weight 691.86
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC140258

 
lab chem
LC140258
Each for $82.17
Description & Specifications

Specifications

Formula Weight 691.86
CAS Max % 100
Health Hazard 1 Warning
Identification Passes Test
Color Reddish
Recommended Storage Room Temperature
Physical Form Powder
Linear Formula C20H6Br4O5·2Na
CAS 17372-87-1
Health Hazard 2 GHS H Statement
Causes serious eye irritation.
Solubility Soluble in water
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Eosin Y
Packaging Amber Glass
Purity Grade Reagent Grade
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes serious eye irritation.

Room Temperature