missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eosin Y, 1% Solution in Water, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 611815000

Catalog No. AC611815000



Description

Description

Indicator

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Specifications

Specifications

Eosin Y
17372-87-1
MFCD00005040
15, 3658
Pass Test
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
745.904
691.85
0.62
Vapor Pressure: 14mmHg at 20°C
C20H12Br4Na2O8
19, 230
2', 4', 5', 2',4',5',7'-Tetrabromofluorescein, disodium salt, C.I. 45380, Eosin Yellowish
MASXMTNVNASWNH-UHFFFAOYSA-L
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
91886399
EcoSafPak™
500mL

Chemical Identifiers

17372-87-1
745.904
MASXMTNVNASWNH-UHFFFAOYSA-L
91886399
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
C20H12Br4Na2O8
MFCD00005040
2', 4', 5', 2',4',5',7'-Tetrabromofluorescein, disodium salt, C.I. 45380, Eosin Yellowish
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SDS Documents