missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eosin Y, 0.25% Alcoholic, Fisher Chemical

Manufacturer:  Fisher Chemical SE22500GH

Catalog No. SE22-500D




Description

Description

EcoSafPak is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications

Specifications

Eosin Y, Alcoholic
67-63-0,67-56-1,17372-87-1,7732-18-5
Acid Red 87
Glass Bottle
Pass Test
0.8g/cm³
500mL
0.25% Solution
Pass Test
Liquid
0.840 to 0.870 (at 25°C)
40mmHg at 200°C
Green-Orange
Documents