missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EOMES, PE-eFluor 610, clone: Dan11mag, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61487580

 View more versions of this product

Catalog No. 501124825



Description

Description

TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.

PE-eFluor® 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red®). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
Specifications

Specifications

EOMES
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O54839
Eomesodermin homolog; EOMES; TBR2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
Dan11mag
PE-eFluor® 610
Eomes
Liquid
Eomes
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
13813
Documents

For Research Use Only.