Please login to your online account to display your discounted pricing

EOMES Rat anti-Mouse, PE-Cy7, Clone: Dan11mag, eBioscience™

Rat Monoclonal Antibody

$201.55 - $506.05

Specifications

Antigen EOMES
Clone Dan11mag
Host Species Rat
Gene Alias Eomesodermin, TBR2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123375

 
affymetrix
25487580
25μg Each for $201.55

501129158

 
affymetrix
25487582
100μg Each for $506.05
Description & Specifications

Specifications

Antigen EOMES
Clone Dan11mag
Host Species Rat
Gene Alias Eomesodermin, TBR2
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This Dan11mag antibody recognizes Eomesodermin (EOMES), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.