missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EOMES Monoclonal Antibody (Dan11mag), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Thermo Fisher Scientific 25487580

 View more versions of this product

Catalog No. 501123375



Description

Description

Description: This Dan11mag antibody recognizes Eomesodermin (EOMES), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Applications Reported: This Dan11mag antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This Dan11mag antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Buffer Set (cat 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

EOMES Monoclonal antibody specifically detects EOMES in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

EOMES
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O54839
Eomes
Affinity chromatography
RUO
13813
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
Dan11mag
PE-Cyanine7
Eomes
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2
Rat
25 μg
Primary
Mouse
Antibody
IgG2a κ
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.