missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eomes Rat anti-Mouse, PE, Clone: Dan11mag, eBioscience™

Rat Monoclonal Antibody

$182.16 - $456.72

Specifications

Antigen Eomes
Clone Dan11mag
Host Species Rat
Gene Alias Eomesodermin, TBR2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010794 Life Technologies
LS12487582
100μg Each for $456.72
501129316 Life Technologies
LS12487580
25μg Each for $182.16
Specifications

Description

This Dan11mag antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.
Description & Specifications

Specifications

Eomes
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Dan11mag
Eomesodermin, TBR2
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL