missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EOMES, eFluor 660, clone: Dan11mag, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS50487582

 View more versions of this product

Catalog No. 501129093



Description

Description

TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.

Description: This Dan11mag antibody recognizes Eomesodermin (EOMES), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.Applications Reported: This Dan11mag antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This Dan11mag antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Buffer Set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Eomes
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54839
Eomesodermin homolog, EOMES, TBR2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
Dan11mag
eFluor 660
Eomes
Liquid
Eomes
IgG2a, kappa, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
13813
Documents

For Research Use Only.