missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EOMES Rat anti-Human, Mouse, Clone: 21Mags8, eBioscience™

Rat Monoclonal Antibody

$143.88 - $302.28

Specifications

Antigen EOMES
Clone 21Mags8
Host Species Rat
Gene Alias Eomesodermin, TBR2
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013075 Life Technologies
LS14487680
25μg Each for $143.88
5013076 Life Technologies
LS14487682
100μg Each for $302.28
Specifications

Description

This 21Mags8 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.

Note: For detection of mouse Eomes, we recommend 12-4875 and 51-4875 and for detection of human Eomes, we recommend 51-4877 due to the brighter staining intensity of these products compared to clone 21Mags8.
Description & Specifications

Specifications

EOMES
Rat
Human, Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
21Mags8
Eomesodermin, TBR2
Flow Cytometry,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL