missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EOMES Mouse anti-Human, PerCP-eFluor 710, Clone: WD1928, eBioscience™

Mouse Monoclonal Antibody

$234.90 - $575.65

Specifications

Antigen EOMES
Clone WD1928
Host Species Murine
Gene Alias Eomesodermin, TBR2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123816 Life Technologies
LS46487742
100 Tests Each for $575.65
501129071 Life Technologies
LS46487741
25 Tests Each for $234.90
Specifications

Description

This WD1928 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.
Description & Specifications

Specifications

EOMES
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
WD1928
Eomesodermin, TBR2
Flow Cytometry (Intracellular Staining)
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test