Learn More
EOMES Monoclonal Antibody (WD1928), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$221.00 - $550.00
Specifications
Antigen | EOMES |
---|---|
Clone | WD1928 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: This WD1928 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Applications Reported: This WD1928 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This WD1928 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Buffer Set (cat. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.Specifications
EOMES | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
Eomes | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Eomes |
WD1928 | |
Flow Cytometry | |
FITC | |
Mouse | |
Human, Porcine | |
O95936 | |
100622125, 8320 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.