Please login to your online account to display your discounted pricing

Eomes Mouse anti-Human, APC-eFluor 780, Clone: WD1928, eBioscience™

Mouse Monoclonal Antibody

$230.55 - $564.05

Specifications

Antigen Eomes
Clone WD1928
Host Species Mouse
Gene Alias Eomesodermin, TBR2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124056

 
life technologies
LS47487742
100 tests Each for $564.05

501124055

 
life technologies
LS47487741
25 tests Each for $230.55
Description & Specifications

Specifications

Antigen Eomes
Clone WD1928
Host Species Mouse
Gene Alias Eomesodermin, TBR2
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

This WD1928 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.