Please login to your online account to display your discounted pricing

EO 1428, Tocris

Selective inhibitor of p38α and p38β2

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material EO 1428
Assay Percent Range >98%
Molecular Formula C20H16BrClN2O
Formula Weight 415.71
Synonym (2-Methylphenyl)-[4-[(2-amino-4-bromophenyl)amino]-2-chlorophenyl]methanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

290810

 
r & d systems
RDS290810
10mg Each for $252.39

290850

 
r & d systems
RDS290850
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 415.71
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C20H16BrClN2O
CAS 321351-00-2
Synonym (2-Methylphenyl)-[4-[(2-amino-4-bromophenyl)amino]-2-chlorophenyl]methanone
Recommended Storage Store at +4°C
Chemical Name or Material EO 1428
Safety and Handling

Store at +4°C