Please login to your online account to display your discounted pricing

Enzyme storage buffer in PBS and glycerol, autoclaved, Alfa Aesar™

For frozen storage of enzymes.

Chemical Identifiers

Chemical Name or Material Enzyme storage buffer in PBS and glycerol
Name Note autoclaved
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAJ63115EQE

 
alfa aesar
J63115EQE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 125mL
Chemical Name or Material Enzyme storage buffer in PBS and glycerol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Name Note autoclaved
Physical Form Liquid
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.