Please login to your online account to display your discounted pricing

Environmental Express™ TCLP Filters


Manufacturer: environmental express  FG77047MM

 View more versions of this product

Catalog No. 09-732-72

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Diameter (English) Outside 1.8 in.
Filter Type Prewashed TCLP filter
For Use With (Equipment) 47mm filter holders
Temperature (English) Maximum 221°F
Temperature (Metric) Max 105°C
Diameter (Metric) Outside 4.7cm
For Use With (Application) Eliminate reagent and waste disposal costs, may reduce blank variability and provide lower levels of background metals compared to onsite washing
Material Borosilicate Glass Fiber