Please login to your online account to display your discounted pricing
Environmental Express™ ProWeigh™ Preweighed Filters for Gravimetric Analysis
Eliminate filter washing, drying, desiccating and preweighing
Manufacturer: environmental express  F93476MM
 View more versions of this product
Catalog No. 09-735-105
  • / Pack of 100 Each
Description & Specifications
Specifications
Product Type ProWeigh Filter for TSS
Weight (English) 2.20 lb.
Weight (Metric) 0.1mg
Size (English) 2.99 in.
Accuracy ±0.4mg
For Use With (Application) Total suspended solids and other gravimetric analyses
Material Glass Fiber