Please login to your online account to display your discounted pricing
Environmental Express™ ProWeigh™ Preweighed Filters for Gravimetric Analysis
Eliminate filter washing, drying, desiccating and preweighing
Manufacturer: environmental express  F93447MM
 View more versions of this product
Catalog No. 09-735-56
  • / Pack of 100 Each
Description & Specifications
Specifications
Accuracy ±0.4mg
Weight (Metric) 0.1mg
Weight (English) 2.20 lb.
Material Glass Fiber
Product Type ProWeigh Preweighed Filter for TSS
Size (English) 1.85 in.
For Use With (Application) Total suspended solids and other gravimetric analyses