H-B Instrument™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers
Certified environmentally safe thermometers replace dangerous mercury.
Manufacturer:  H-B Instrument™ B605030100
 View more versions of this product
Catalog No. S95814
Description
Description
  • Earth friendly, EnviroKleen™ certified thermometers
  • Feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass
  • Meet government, education, and industry mandates to purchase “Green” products
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Order Info
Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Compliance
Manufactured and tested by USA quality team.
Specifications
Specifications
2°F
500°F
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
76mm
14 in.
±3°F, ±4°F above 230°F, ±5°F above 392°F
20° to 500°F
20°F
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Partial Immersion
350mm
Documents