Please login to your online account to display your discounted pricing

Alfa Aesar™ Enterostatin, Human

For cellular and molecular biology applications

Specifications

Product Type Enterostatin, human
Cross Reactivity Human
Molecular Weight 572.69
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Gly-Pro-Arg
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66199MA

 
alfa aesar™
J66199MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66199MB

 
alfa aesar™
J66199MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Enterostatin, human
Cross Reactivity Human
Molecular Weight 572.69
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Gly-Pro-Arg
Format Solid
Regulatory Status RUO
Purification Method HPLC