MilliporeSigma™ Entellan™ New; For Microscopy
Mounts medium for cover slippers
Manufacturer:  MilliporeSigma™ 1079610100
 View more versions of this product
Catalog No. M1079610100
Description
Description
  • Suited for commercial automated mounting instruments that operate with cover slippers
  • Viscosity: 500 to 600mPas
  • Narrow viscosity range improves accelerates the application
  • No need to reset after first run
Specifications
Specifications
Glass Bottle
15° to 25°C
100mL