missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Entecavir Monohydrate, TCI America™

Manufacturer:  TCI America E0899200MG

 View more versions of this product

Catalog No. E0899200MG



Specifications

Specifications

Entecavir Monohydrate
259°C
209216-23-9
MFCD09754448
YXPVEXCTPGULBZ-WQYNNSOESA-N
2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one;hydrate
16052026
277.28
≥98.0% (HPLC,T)
White
200mg
C12H17N5O4
entecavir hydrate, entecavir monohydrate, entecavir monohydrate, baraclude tn, entecavir usan, entecavir hydrate jan, baraclude, 9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylenecyclopentyl guanine monohydrate, entecavir inn, 2-amino-9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylidenecyclopentyl-1,9-dihydro-6h-purin-6-one-water 1/1
C=C1C(CC(C1CO)O)N2C=NC3=C2NC(=NC3=O)N.O
295.299
CHEBI:59902
Crystal-Powder at 20°C

Chemical Identifiers

209216-23-9
295.299
YXPVEXCTPGULBZ-WQYNNSOESA-N
16052026
2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one;hydrate
C12H17N5O4
MFCD09754448
entecavir hydrate, entecavir monohydrate, entecavir monohydrate, baraclude tn, entecavir usan, entecavir hydrate jan, baraclude, 9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylenecyclopentyl guanine monohydrate, entecavir inn, 2-amino-9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylidenecyclopentyl-1,9-dihydro-6h-purin-6-one-water 1/1
CHEBI:59902
C=C1C(CC(C1CO)O)N2C=NC3=C2NC(=NC3=O)N.O
Safety and Handling

Safety and Handling

TSCA : No

Documents