Please login to your online account to display your discounted pricing

Enoximone, Tocris

PDE3 inhibitor

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material Enoximone
Assay Percent Range >99%
Molecular Formula C12H12N2O2S
Formula Weight 248.3
Synonym 1, 3-Dihydro-4-methyl-5-[4-(methylthio)benzoyl]-2H-imidazol-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

169350

 
r & d systems
RDS169350
50mg Each for $816.90

169310

 
r & d systems
RDS169310
10mg Each for $195.49
Description & Specifications

Specifications

Formula Weight 248.3
Quantity 50mg
Assay Percent Range >99%
Molecular Formula C12H12N2O2S
Synonym 1, 3-Dihydro-4-methyl-5-[4-(methylthio)benzoyl]-2H-imidazol-2-one
CAS 77671-31-9
Chemical Name or Material Enoximone
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C