English/Metric Electronic Digital Caliper
Manufacturer:  Marathon Watch Company 030300
 View more versions of this product
Catalog No. S40503-4D
Specifications Description & Specifications
Specifications
Range: 0 to 300mm
Large LCD screen display, carrying ABS shock-resistant case and extra battery
Electronic digital caliper
Offers metric/inch conversion, origin setting, data hold, relative/absolute measuring and tolerance setting and to measure the distance between two symmetrically opposing sides