English/Metric Electronic Digital Caliper
Manufacturer:  Marathon Watch Company 030150
 View more versions of this product
Catalog No. S40503-2D
Specifications Description & Specifications
Specifications
Electronic digital caliper
Large LCD screen display, carrying ABS shock-resistant case and extra battery
Range: 0 to 150mm
Offers metric/inch conversion, origin setting, data hold, relative/absolute measuring and tolerance setting and to measure the distance between two symmetrically opposing sides