Please login to your online account to display your discounted pricing

Alfa Aesar™ Endothelin-2, Human

For cellular and molecular biology applications

Specifications

Product Type Endothelin-2, human
Molecular Weight 2546.91
Storage Requirements -30° to -10°C
Sequence Cys-Ser-Cys-Ser-Ser-Trp-Leu-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66594EXD

 
alfa aesar™
J66594EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66594MCR

 
alfa aesar™
J66594MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Endothelin-2, human
Molecular Weight 2546.91
Storage Requirements -30° to -10°C
Sequence Cys-Ser-Cys-Ser-Ser-Trp-Leu-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH
Format Solid
Regulatory Status RUO
Purification Method HPLC