Please login to your online account to display your discounted pricing

Alfa Aesar™ Endothelin-1 (1-15)Amide, Human

For cellular and molecular biology applications

Specifications

Product Type Endothelin-1 (1-15) amide, human
Cross Reactivity Human
Molecular Weight 1717.04
Storage Requirements -30° to -10°C
Sequence Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66107LB0

 
alfa aesar™
J66107LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66107MA

 
alfa aesar™
J66107MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Endothelin-1 (1-15) amide, human
Cross Reactivity Human
Molecular Weight 1717.04
Storage Requirements -30° to -10°C
Sequence Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC