Please login to your online account to display your discounted pricing

Alfa Aesar™ Endothelial Nitric Oxide Synthase (eNOS)(205-215), Certified Mass Spec Grade

For cellular and molecular biology applications

Specifications

Product Type Endothelial Nitric Oxide Synthase (eNOS) (205-215), Certified Mass Spec Grade
Molecular Weight 1398.60
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence EMFTYICNHIK
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67806LPL

 
alfa aesar™
J67806LPL
1 vial, 500 pmol CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67806ODL

 
alfa aesar™
J67806ODL
3 vials, 500 pmol/vial CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Endothelial Nitric Oxide Synthase (eNOS) (205-215), Certified Mass Spec Grade
Molecular Weight 1398.60
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence EMFTYICNHIK
Regulatory Status RUO
Purification Method HPLC