Please login to your online account to display your discounted pricing

Alfa Aesar™ Endothelial Nitric Oxide Synthase (eNOS)(194-204), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67775LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67775LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1 vial, 500 pmol
Format Lyophilized powder
Sequence VFDARDCRSAQ
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Endothelial Nitric Oxide Synthase (eNOS) (194-204), Certified Mass Spec Grade
Molecular Weight 1267.40
Purification Method HPLC