Please login to your online account to display your discounted pricing

Endothelial Cell Growth Suppl. (Lyoph. Powd. W/ Streptomycin Sulfate), Fisher BioReagents


Manufacturer: fisher scientific company  BP265450

 View more versions of this product

Catalog No. BP265450

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Specifications

Packaging Amber glass
Product Type Endothelial Cell Growth Supplement
Quantity 50mg
Storage Requirements 2° to 8°C
Format Powder
Quantity 50mg