missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Endomucin, eFluor 660, clone: eBioV.7C7 (V.7C7), eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS50585180

 View more versions of this product

Catalog No. 501129166



Description

Description

EMCN is a mucin-like sialoglycoprotein that interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.

Description: The eBioV.7C7 monoclonal antibody reacts with mouse endomucin, which was identified in a search for cell-surface expressed endothelial cell markers. Endomucin is a 75kDa type I integral membrane protein, with similarities to the sialomucin family of proteins including extensive O-linked glycosylation. Endomucin is expressed on endothelial cells, however, an exception is the high endothelial venules (HEV) of secondary lymphoid organs. In addition, it has been demonstrated that endomucin is expressed on CD34-c-Kit+Sca-1+Lin- hematopoietic progenitors, and that these cells are capable of multi-lineage long-term reconstitution of the hematopoietic compartment.Applications Reported: This eBioV.7C7 (V.7C7) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioV.7C7 (V.7C7) antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Endomucin
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9R0H2
EMCN2, MUC14, MUC-14, endomucin-2, gastric cancer antigen Ga34, mucin-14
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBioV.7C7 (V.7C7)
eFluor 660
Emcn
Liquid
Emcn
IgG2a
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
59308
Documents

For Research Use Only.