missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Endomorphin-2
$126.05 - $486.53
Specifications
CAS | 141801-26-5 |
---|---|
Molecular Formula | C32H37N5O5 |
Molecular Weight (g/mol) | 571.678 |
MDL Number | MFCD01321064 |
InChI Key | XIJHWXXXIMEHKW-LJWNLINESA-N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ66953LB0 |
Alfa Aesar™
J66953LB0 |
5mg |
Each for $126.05
|
|
AAJ66953MB |
Alfa Aesar™
J66953MB |
25mg |
Each for $486.53
|
|
Specifications
141801-26-5 | |
571.678 | |
XIJHWXXXIMEHKW-LJWNLINESA-N | |
5311081 | |
C1CC(N(C1)C(=O)C(CC2=CC=C(C=C2)O)N)C(=O)NC(CC3=CC=CC=C3)C(=O)NC(CC4=CC=CC=C4)C(=O)N |
C32H37N5O5 | |
MFCD01321064 | |
2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl-n-2s-1-2s-1-amino-1-oxo-3-phenylpropan-2-yl amino-1-oxo-3-phenylpropan-2-yl pyrrolidine-2-carboxamide | |
(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]-N-[(2S)-1-[[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]amino]-1-oxo-3-phenylpropan-2-yl]pyrrolidine-2-carboxamide |