missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Endomorphin-1

$126.59 - $488.85

Chemical Identifiers

CAS 189388-22-5
Molecular Formula C34H38N6O5
Molecular Weight (g/mol) 610.715
MDL Number MFCD01321063
InChI Key ZEXLJFNSKAHNFH-SYKYGTKKSA-N
Synonym 2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl-n-2s-1-2s-1-amino-1-oxo-3-phenylpropan-2-yl amino-3-1h-indol-3-yl-1-oxopropan-2-yl pyrrolidine-2-carboxamide
PubChem CID 5311080
IUPAC Name (2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]-N-[(2S)-1-[[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]amino]-3-(1H-indol-3-yl)-1-oxopropan-2-yl]pyrrolidine-2-carboxamide
SMILES C1CC(N(C1)C(=O)C(CC2=CC=C(C=C2)O)N)C(=O)NC(CC3=CNC4=CC=CC=C43)C(=O)NC(CC5=CC=CC=C5)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66775LB0 View Documents Alfa Aesar™
J66775LB0
5mg Each for $126.59
 
AAJ66775MB View Documents Alfa Aesar™
J66775MB
25mg Each for $488.85
 
Specifications

Chemical Identifiers

189388-22-5
610.715
ZEXLJFNSKAHNFH-SYKYGTKKSA-N
5311080
C1CC(N(C1)C(=O)C(CC2=CC=C(C=C2)O)N)C(=O)NC(CC3=CNC4=CC=CC=C43)C(=O)NC(CC5=CC=CC=C5)C(=O)N
C34H38N6O5
MFCD01321063
2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl-n-2s-1-2s-1-amino-1-oxo-3-phenylpropan-2-yl amino-3-1h-indol-3-yl-1-oxopropan-2-yl pyrrolidine-2-carboxamide
(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]-N-[(2S)-1-[[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]amino]-3-(1H-indol-3-yl)-1-oxopropan-2-yl]pyrrolidine-2-carboxamide

Specifications

White
189388-22-5
MFCD01321063
ZEXLJFNSKAHNFH-SYKYGTKKSA-N
(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]-N-[(2S)-1-[[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]amino]-3-(1H-indol-3-yl)-1-oxopropan-2-yl]pyrrolidine-2-carboxamide
5311080
Solid
5mg
C34H38N6O5
2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl-n-2s-1-2s-1-amino-1-oxo-3-phenylpropan-2-yl amino-3-1h-indol-3-yl-1-oxopropan-2-yl pyrrolidine-2-carboxamide
C1CC(N(C1)C(=O)C(CC2=CC=C(C=C2)O)N)C(=O)NC(CC3=CNC4=CC=CC=C43)C(=O)NC(CC5=CC=CC=C5)C(=O)N
610.715
610.7
Endomorphin-1
Safety and Handling

Safety and Handling

RTECSNumber : SQ7316800

storageNote1 : -30° to -10°C