Please login to your online account to display your discounted pricing

Alfa Aesar™ Endomorphin-1


Quantity 25mg

Manufacturer: alfa aesar™  J66775MB

 View more versions of this product

Catalog No. AAJ66775MB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 610.7
Chemical Name or Material Endomorphin-1
CAS 189388-22-5
Physical Form Solid
MDL Number MFCD01321063
Synonym Tyr-Pro-Trp-Phe-NH2
Storage Note 1 -30° to -10°C
Color White to off-white
Quantity 25mg
RTECS Number SQ7316800

Specifications

Chemical Name or Material Endomorphin-1
CAS 189388-22-5
Formula Weight 610.7
MDL Number MFCD01321063
Synonym Tyr-Pro-Trp-Phe-NH2
Accessories

-30° to -10°C