Please login to your online account to display your discounted pricing

Alfa Aesar™ Endokinin C, Human

For cellular and molecular biology applications

Specifications

Product Type Endokinin C, human
Cross Reactivity Human
Molecular Weight 1674.94
Storage Requirements -30° to -10°C
Sequence Lys-Lys-Ala-Tyr-Gln-Leu-Glu-His-Thr-Phe-Gln-Gly-Leu-Leu-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66600LB0

 
alfa aesar™
J66600LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66600MA

 
alfa aesar™
J66600MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Endokinin C, human
Cross Reactivity Human
Molecular Weight 1674.94
Storage Requirements -30° to -10°C
Sequence Lys-Lys-Ala-Tyr-Gln-Leu-Glu-His-Thr-Phe-Gln-Gly-Leu-Leu-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC