Please login to your online account to display your discounted pricing

endo-IWR 1, Tocris™

Axin stabilizer; promotes β-catenin phosphorylation

$179.63 - $750.85

Chemical Identifiers

Chemical Name or Material endo-IWR 1
Assay Percent Range >98%
Molecular Formula C25H19N3O3
Formula Weight 409.44
Synonym [(3aR™,4S™,7R™,7aS)-1,3,3a,4,7,7a-Hexahydro-1,3-dioxo-4,7-methano-2H-isoindol-2-yl]-N-8-quinolinylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

353210

 
r & d systems
RDS353210
10mg Each for $179.63

353250

 
r & d systems
RDS353250
50mg Each for $750.85
Description & Specifications

Specifications

Formula Weight 409.44
Quantity 10mg
Chemical Name or Material endo-IWR 1
Assay Percent Range >98%
Molecular Formula C25H19N3O3
Synonym [(3aR™,4S™,7R™,7aS)-1,3,3a,4,7,7a-Hexahydro-1,3-dioxo-4,7-methano-2H-isoindol-2-yl]-N-8-quinolinylbenzamide
CAS 1127442-82-3
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature