Please login to your online account to display your discounted pricing

endo-3-Bromo-D-camphor, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material endo-3-Bromo-D-camphor
Name Note 99+%
CAS 10293-06-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H15BrO
Formula Weight 231.13
MDL Number MFCD00003744
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC164225000

  SDS
thermo fisher scientific
164225000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Needles or Crystalline Powder
Assay ≥99.0%
Formula Weight 231.13
Quantity 500g
Color White
Beilstein 07, II, 101
Merck Index 09, 1414
EINECS Number 233-652-1
Melting Point 74° to 78°C
RTECS Number ED6750000
MDL Number MFCD00003744
Infrared Spectrum Authentic
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material endo-3-Bromo-D-camphor
Storage Information May discolor on exposure to light
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C10H15BrO
CAS 10293-06-8
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May be corrosive to metals.
Specific Rotation +128° to +132° (20°C, 589nm) (c=5, CH3OH)
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May be corrosive to metals.