Endicott-Seymour Foster Caddy Vial Rack
Manufacturer:  Endicott-Seymour 1015*
Catalog No. 03-337-80
Description
Description
  • Made of high-density polypropylene
  • Autoclavable
  • Chemical- and heat-resistant
  • Holds up to 150 17mm dia. miniature vials
Specifications
Specifications
17mm dia. Miniature liquid scintillation counting vials
4.8 cm
32.4 cm
150
1.87 in.
12.75 in.
HDPE
Documents