missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Encon™ Yello-Bowl™ Pedestal-Mount Fountain Eyewashes
Provides first-aid treatment for hazardous material exposure
$585.71 - $585.71
Specifications
Flow Rate | 3.2 gpm |
---|---|
Mounting | Pedestal |
Pressure Range | 30 to 70psi |
Product Type | Pedestal mount fountain |
Includes: An emergency sign of size 7 x 11 in. (17.8 x 28cm) with legend “EMERGENCY EYEWASH”.
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
29-663-183
|
Encon™
01030302 |
ABS Plastic dual stream head, wye, Float-off covers, Stainless Steel bead chains, ABS Plastic yellow bowl, self adjusting regulator, supply connection, emergency sign |
Each for $585.71
|
|
Description
- ABS plastic heads, wye and float off covers are secured with SS bead chains
- High visibility ABS plastic, 10.8 in. dia. (27.4cm) Yello-Bowl
- Self-adjusting regulator to flow at 3.2 gpm (12.1L/min.)
- Supply connection: 0.5 in. NPT
- Waste connection: 1.25 in. NPT
- Model 01030305 comes with a foot treadle
Specifications
3.2 gpm | |
Pedestal | |
30 to 70psi | |
Pedestal mount fountain |