missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Encon™ Yello-Bowl™ Pedestal-Mount Fountain Eyewashes
Provides first-aid treatment for hazardous material exposure
Supplier: Encon™ 01030302
Includes: An emergency sign of size 7 x 11 in. (17.8 x 28cm) with legend “EMERGENCY EYEWASH”.
Description
- ABS plastic heads, wye and float off covers are secured with SS bead chains
- High visibility ABS plastic, 10.8 in. dia. (27.4cm) Yello-Bowl
- Self-adjusting regulator to flow at 3.2 gpm (12.1L/min.)
- Supply connection: 0.5 in. NPT
- Waste connection: 1.25 in. NPT
- Model 01030305 comes with a foot treadle
Specifications
3.2 gpm | |
Pedestal | |
Pedestal Mount Fountain |
ABS Plastic Dual Stream Head, wye, ABS Plastic Float-off Covers, BCP wye, Self Adjusting Regulator, Stainless Steel Bead Chains, Stainless Steel Valve Stem, Actuation Graphic | |
30 to 70 psi |