Please login to your online account to display your discounted pricing

Encon™ Wall-Mount Eye Wash

For emergency use, outfit your workspace with the Encon Wall-Mount Eye Wash.

$447.24 - $634.74

Specifications

Certifications/Compliance ANSI Z358.1-2004
For Use With (Application) First aid treatment for hazardous material exposure
Pressure Range 30 to 70psi
View More Specs
Products
Catalog Number Mfr. No. Includes Mounting Product Type Price Quantity    

18-999-339E

 
encon™
01045001
Acetal eye/facewash head, BCP wye, Plastic hinge cover, Stainless Steel bowl Wall Wall mount fountain Each for $634.74

19-021702

 
encon™
01030401
High visibility bowl, ABS Plastic head, wye and Float-off covers, Stainless Steel bead chains, self adjusting regulator, Chrome Plated Brass valve Wall Eyewash Bottle Each for $447.24
Description & Specifications

Specifications

Certifications/Compliance ANSI Z358.1-2004
For Use With (Application) First aid treatment for hazardous material exposure
Pressure Range 30 to 70psi

  • Can be configured with either an ABS Yello-Bowl™ or SS Bowl
  • Piping is configurable with SS, Galvanized or PVC
 
Piping, Valve and Pedistal Choices
  • BCP, PVC and SS Piping 1.27cm (0.5 in.)
  • CP bronze stay-open ball valve with CP ball; SS valve stem; SS push plate with actuation graphic 1.27cm (0.5 in.)
  • Schedule 80 galvanized steel, PVC and SS pedestal
 
Eyewash Choices
  • ABS plastic heads, wye and float off covers secured with SS bead chans; self adjusting regulator to flow at 3.2 gpm (12.1 lpm)
  • Acetal heads with hinged covers, BCP wye; self-adjusting regulation to flow at 2.4 gpm (9.1 lpm)
  • SS heads with hinged covers, SS wye; self-adjusting regulator to flow at 2.5 gpm (9.5 lpm) (available only with SS piping and valves)
 
Facewash Choices
  • ABS plastic heads, wye and float off covers secured with SS bead chans; self adjusting regulator to flow at 8.0 gpm (30.3 lpm)
  • ABS plastic heads and float off covers secured with SS bead chains, BCP wye; self-adjusting regulator to flow at 8.0 gpm (30.3 lpm)
 
Bowl
  • ABS plastic Yello-Bowl™ 27.4cm (10.8 in.) diameter
  • 304 SS bowl, 26.9cm (10.6 in.) diameter
  •  
Bowl Cover Options
  • Corrosion-resistant high visibility ABS plastic; high visibility
  • SS push plate actuator opens cover
  • Manually closing cover is independent of water flow actuator
  • 304 SS; push plate actuator opens cover
  • Available on models with bowls and BCP wyes only
 
Bowl Waste Options
  • 3.17 cm x 15.24cm (1.25 x 6 in.) slip joint tailpiece
  • 3.17cm (1.25 in.) NPT Connection.
  • P-trap - 3.17cm (1.25 in.) tube connection