missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Encon™ Supply Pipe-Mount Facewash

Self-adjusting regulator which provides 8.0 gpm (30.3Lpm) flow

Supplier:  Encon™ 01035601

 View more versions of this product

Catalog No. 29-663-187



Description

Description

  • ABS plastic heads
  • Wye are available either in ABS plastic or Brass chrome-plated material
  • Float off covers secured with Stainless steel bead chains
  • Can be configured with either an ABS Yello-Bowl™, SS Bowl, or No Bowl
  • Piping is configurable with Stainless steel, Galvanized or PVC
  • 0.5 in. NPT connection

Certifications

All facewashes meet the ANSI Z358.1 requirements of an eyewash.

Specifications

Specifications

ANSI Z358.1
ABS Plastic Dual Stream Head, wye, ABS Plastic Float-off Covers, BCP wye, Self Adjusting Regulator, Stainless Steel Bead Chains, Stainless Steel Valve Stem, Actuation Graphic
Pipe
30 to 70 psi
8 gpm
Without bowl
1.25 in. NPT
Mount Fountain
SDS
Documents

Documents

Product Certifications
Promotions

Promotions