Please login to your online account to display your discounted pricing
Encon™ Shower/Eyewash Signs
$193.28 - $239.07
Specifications
Type Shower/eyewash sign
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
18-999-339B
encon safety products
01112953
Green Each for $239.07
19-021789
encon safety products
01112911
Yellow Each for $193.28
Description & Specifications
Specifications
Type Shower/eyewash sign
Quantity Each
High-visibility graphics on a bright, chemical-resistant yellow or green ABS plastic shell promote awareness of emergency shower/eyewash equipment.
Signs mount on a top/bottom entry shower. Include tie wraps and trim strip. Can also be attached by means of wall screws, rivets, or tape.