Please login to your online account to display your discounted pricing
Encon Countertop Eyewashes
$755.18 - $757.43
Specifications
Activation Stainless Steel push plate
Certifications/Compliance ANSI Z358.1-1998
Flow Rate 2.4 gpm
For Use With (Application) Laboratory use
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    
18-999-341B
encon safety products
01040123
Acetal eyewash head, hinged cover, vertical valve, right hand actuation, self adjusting regulator, Chrome Plated Bronze ball valve and ball Each for $757.43
18-999-341C
encon safety products
01040124
Acetal eyewash head, hinged cover, vertical valve, left hand actuation, self adjusting regulator, Chrome Plated Bronze ball valve and ball Each for $755.18
Description & Specifications
Specifications
Activation Stainless Steel push plate
Certifications/Compliance ANSI Z358.1-1998
Flow Rate 2.4 gpm
For Use With (Application) Laboratory use
Swivel and Saf-T-Lok ™ assemblies allow eyewash to swing 90° so it is over the sink; then it locks into ready-to-use position. Following use, just lift top portion of Saf-T-Lok ™ assembly to restore station to storage position. Station features chrome-plated bronze stay-open ball valve with stainless-steel push plate and graphic. Dual eyewash heads made of high visibility ABS plastic. Float-off covers release automatically when water is turned on. Self-adjusting regulators provide constant, even flow of 3.5 gpm (13.2L/min.) under varying hydraulic conditions. Piping, fittings, and swivel are of 3/8 in. (1cm) chrome-plated brass. Vandal-resistant base connects through countertop. With right- or left-hand push plate.
Meet ANSI Z358.1-1998.