missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Encon™ Combination Shower/Eyewash Units

Encon Combination Shower/Eyewash Unit provides workers with quick access to eye flushing and showering during an emergency situation.

Supplier:  Encon™ 01050223

 View more versions of this product

Catalog No. 29-663-163



Description

Description

  • Floor mounted/wall supported shower and eyewash station
  • Features heavy-wall Schedule 80 corrosion-resistant PVC piping and socket weld joints
  • Shower operated by stainless-steel pull rod that bears bright yellow actuation graphic
  • Yellow ABS plastic shower head delivers a minimum 51cm (20 in.) dia. pattern to target area 152cm (60 in.) above standing level
  • Eyewash features yellow ABS plastic dual stream heads and float-off covers secured by stainless-steel bead chain
  • Activated by stainless-steel push plate with actuation graphic
  • Self-adjusting 3.5 gpm (13.2L/min.) regulator assures a constant, even flow under varying hydraulic conditions
  • With ABS Yello-Bowl receptacle bowl
  • Stay-open 1 1/2 in. (3.8cm) shower valve and 1/2 in. (1.3cm) eyewash valve are of polane-coated bronze
  • Ball is chrome-plated

Order Info

Supplied with 7W × 11 in. H (18 × 28cm)

Certifications

CSA Z94.3-2007 standards.

Compliance

Meet ANSI Z358.1-2009

Specifications

Specifications

Push Plate and Pull Rod
3.2 to 30 gpm
Floor
30 to 70 psi
ANSI Z358.1-2009, CSA Z94.3-2007
Yellow-bowl, type 304 schedule 40 Stainless Steel piping, open ball valves, yellow ABS Plastic shower head, ABS Plastic dual stream Eyewash head, wye
1.25 in. NPT
Combination Shower, Eyewash Unit
SDS
Documents

Documents

Promotions

Promotions