missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Enameled Magnet Wire

Use to construct electrical studies.

Manufacturer:  Fisher Science Education WS0149

 View more versions of this product

Catalog No. S4828B



Description

Description

Insulated.

For constructing or repairing:

  • Chokes
  • Dynamos
  • Generators
  • Motors
  • Electronic coils
  • Transformers
  • Transmitters
  • Tuners
  • Solenoids

Order Info

Comes in 1 lb. spool (454g).

Specifications

Specifications

Constructing or repairing chokes, dynamos, generators, motors, electronic coils, transformers, transmitters, tuners, solenoids
Copper-enameled Magnet Wire
24 ga
243.84 m
0.45 kg
Copper
Construct electrical studies
800 ft.
1 lb.
SDS
Documents
Product Certifications
Promotions

Promotions