missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Emtricitabine, TCI America™

Manufacturer:  TCI America E1007250MG

 View more versions of this product

Catalog No. E1007250MG



Specifications

Specifications

Emtricitabine
155°C
143491-57-0
MFCD00870151
C1C(OC(S1)CO)N2C=C(C(=NC2=O)N)F
247.244
CHEBI:31536
Crystal-Powder at 20°C
White-Yellow
250mg
C8H10FN3O3S
XQSPYNMVSIKCOC-NTSWFWBYSA-N
4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
60877
247.24
≥98.0% (HPLC,T)

Chemical Identifiers

143491-57-0
247.244
XQSPYNMVSIKCOC-NTSWFWBYSA-N
CHEBI:31536
C1C(OC(S1)CO)N2C=C(C(=NC2=O)N)F
C8H10FN3O3S
MFCD00870151
60877
4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
Safety and Handling

Safety and Handling

RTECSNumber : UW7360500

TSCA : No

Documents